Centrifugebuizen, conische bodem, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3103-0015 3103-0050
525-0464EA 47.5 EUR
525-0464 525-0465
Centrifugebuizen, conische bodem, Nalgene®
Buisjes Centrifugebuisjes
PPCO, translucent.
  • Excellent chemical resistance
  • Maximum RCF: 6000 ×g
  • Autoclavable
Conical-bottom centrifuge tubes, which feature molded-in graduations.