Flessen, brede hals, HDPE, rond, met schroefdop, IP2
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
312199-0004 312199-0008 312199-0016 312199-0032
NALG312199-0004EA 1640 EUR
NALG312199-0004 NALG312199-0008 NALG312199-0016 NALG312199-0032
Flessen, brede hals, HDPE, rond, met schroefdop, IP2
Flessen Kunststof flessen
Wide-mouth HDPE IP 2 bottles, translucent with PP screw cap.
  • Good chemical resistance
  • Heat resistant up to +120 °C
  • Leakproof when used with Nalgene closures
  • Round shape
Safe, durable, economical bottles for the transport and packaging of hazardous goods. Recommended for customers who wish to put together and certify their own packaging system. Extra-wide mouth for simple filling and emptying of liquids and dry substances. The sturdy walls withstand cracking and damage from impact caused by shocks during transport. The bottom of the bottle incorporates IP2 marking, material, volume and registration notch. Evaluated at 15 psi per 49 CFR 173.27 © (2), ICAO Technical Instructions Part 4; 1.1.6 and IATA Dangerous Goods Regulations, Section 5.0.2.9.
Verpakking: Bottles and closures are bulk packed in separate polybags within the same carton.