Mierenzuur 100% 49 - 51% (by titrimetric analysis) in water, Puriss. p.a. for HPLC, for HPLC, Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Aminic Acid, Methanoic acid
09676-100ML 09676-500ML
09676-100MLEA 65.3 EUR
09676-100ML 09676-500ML
Mierenzuur 100% 49 - 51% (by titrimetric analysis) in water, Puriss. p.a. for HPLC, for HPLC, Fluka™
Mierenzuur
Formule: HCO₂H
Kookpunt: 101 °C (1,013 hPa. Anhydrous substance)
Smeltpunt: 4 °C (anhydr.)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003297
CAS nummer: 64-18-6
EINECS: 200-579-1
UN: 3412
ADR: 8,II
Bereik: 01-2119491174-37-XXXX
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=a77e0bed84455e37172a184511237ad1d178fd11&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay 49.0 - 51.0 %
Absorbance at 270 nm Max. 0.05
Absorbance 280 nm Max. 0.04
Absorbance at 290 nm Max. 0.03
Absorbance at 300 nm Max. 0.02
Absorbance 500 nm Max. 0.02