Spuitfilters geoptimaliseerd voor schaal omhoog, Acrodisc®
Leverancier: Cytiva (Formerly Pall Lab)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4902-N 4905-N 4906-N 4907-N 4908-N
514-4238EA 249 EUR
514-4238 516-8971 516-8972 516-8973 516-7717
Spuitfilters geoptimaliseerd voor schaal omhoog, Acrodisc®
Filters Spuitfilters
Simplifies scale up and minimises revalidation; no need to change membrane materials during transition to pilot or production. PP housing; four membrane chemistries assure compatibility with a wide range of fluids.
  • Integrity testable (water bubble point)
  • Bacterial retention tested
  • Sterilised by gamma irradiation to eliminate potential contamination by ethylene oxide residuals
Supor® membrane has high flow rates and throughputs and is ideal for solutions where low protein binding is required. Not recommended with some ketones.
Fluorodyne® II membrane offers high flow rates and is ideal for applications where PVDF membrane is specified. Not recommended with some ethers.
Ultipor® membrane provides broad solvent and chemical compatibility and low extractables.
Posidyne® membrane enhances bioburden and pyrogen removal from aqueous solutions.
Applications: Drug development studies, determination of product compatibility and recovery, preliminary filterability testing, small-volume liquid sterilisation.