Natriumhydroxide 50% (w/w) in aqueous solution
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
33382.22 33382.A1 33382.A7
33382.22EA 46.7 EUR
33382.22 33382.A1 33382.A7
Natriumhydroxide 50% (w/w) in aqueous solution
Natriumhydroxide
CAS No.: 1310-73-2 Molecular Formula: NaOH Formula Weight: 40.00 Physical Form: Liquid Density: 1.53 Appearance: Colourless UN No.: 1824 MDL No.: MFCD00003548
Formule: NaOH
Kookpunt: 119…135 °C (1013 hPa)
Smeltpunt: 9…10 °C
Dichtheid: 1,53
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003548
CAS nummer: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II