Trechters met korte steel, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4256-0234 4256-0414 4256-0638
NALG4256-0234EA 43.8 EUR
NALG4256-0234 NALG4256-0414 NALG4256-0638
Trechters met korte steel, Nalgene®
Trechters Filtertrechters Filtertrechters
PP or HDPE, translucent.
  • Shatter-proof
  • PP: autoclavable
  • HDPE: not autoclavable
Order Now
Specificaties
Environmentally Preferable Lage impact op de fabricage
Milieu certificering NAR Certificate of Retirement