Slangaansluitingen, T-vormig, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6151-0125 6151-0187 6151-0250 6151-0312 6151-0375 6151-0500
NALG6151-0125EA 31.9 EUR
NALG6151-0125 NALG6151-0187 NALG6151-0250 NALG6151-0312 NALG6151-0375 NALG6151-0500
Slangaansluitingen, T-vormig, Nalgene®
Fittingen en connectoren
PP, translucent.
  • Free of animal derived components
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Autoclavable
Verpakking: Double-bagged to maintain cleanliness.