Slangaansluitingen, Y-vormig, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6152-0125 6152-0187 6152-0250 6152-0312 6152-0375 6152-0500
NALG6152-0125EA 34.5 EUR
NALG6152-0125 NALG6152-0187 NALG6152-0250 NALG6152-0312 NALG6152-0375 NALG6152-0500
Slangaansluitingen, Y-vormig, Nalgene®
Fittingen en connectoren
PP, translucent.
  • Material free of animal derived components
  • Excellent chemical resistance
  • Excellent temperature resistance
Verpakking: Double-bagged to maintain cleanliness.