Potten met deksel, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5352-0002 5352-0004
NALG5352-0002EA 68.6 EUR
NALG5352-0002 NALG5352-0004
Potten met deksel, Nalgene®
Potjes Potjes voor algemeen gebruik
PP, translucent.
  • Straight sides and no neck making access and retrieval easy
  • Larger sizes make excellent waste crocks
  • Autoclavable
Ideal for storage and transport of solids.