Perrhenic acid 75 - 80% ≥75-80%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
11413.06
11413.14
11413.06EA
134
EUR
11413.06
11413.14
Perrhenic acid 75 - 80% ≥75-80%
Perrhenic acid
CAS No.: 13768-11-1
Molecular Formula: HO4Re
Formula Weight: 251.26
Density: 2.42
Appearance: Colourless to pale yellow
UN No.: 3264
MDL No.: MFCD00011326
Formule:
HO₄Re Dichtheid: 2,42 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00011326 CAS nummer: 13768-11-1 EINECS: 237-380-4 UN: 3264 ADR: 8,II |