Osmiumtetraoxide 2% in aqueous solution
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Osmium (VIII) oxide, Osmic acid anhydride, Osmic acid
45384.AB 45384.9W
45384.ABEA 164 EUR
45384.AB 45384.9W
Osmiumtetraoxide 2% in aqueous solution
Osmiumtetraoxide
CAS No.: 20816-12-0 Molecular Formula: OsO4 Formula Weight: 254.10 Physical Form: Liquid Appearance: Colourless to pale yellow Odour: Chlorine-like Merck Reference: 14,6893 UN No.: 3287 MDL No.: MFCD00011150
Formule: O₄Os
MW: 254.23 g/mol
Kookpunt: 130
Smeltpunt: 40
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011150
CAS nummer: 20816-12-0
EINECS: 244-058-7
UN: 3287
ADR: 6.1 III
Merck Index: 14,6893