Osmiumtetraoxide 4% (w/w) in water
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Osmium (VIII) oxide, Osmic acid anhydride, Osmic acid
319010050 319010100
ACRO319010050EA 168 EUR
ACRO319010050 ACRO319010100
Osmiumtetraoxide 4% (w/w) in water
Osmiumtetraoxide
Formule: O₄Os
MW: 254.23 g/mol
Kookpunt: 130
Smeltpunt: 40
Dichtheid: 1.04 g/cm³
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00011150
CAS nummer: 20816-12-0
EINECS: 244-058-7
UN: 3287
ADR: 6.1 II
Merck Index: 13,06962
Order Now
Specification Test Results
Appearance Clear yellow Solution
Strength 3.92 to 4.08 %