Osmiumtetraoxide 4% in aqueous solution
Leverancier: Serva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Osmium (VIII) oxide, Osmic acid anhydride, Osmic acid
31253.01 31253.02 31253.03 31253.04
SERA31253.01EA 40 EUR
SERA31253.01 SERA31253.02 SERA31253.03 SERA31253.04
Osmiumtetraoxide 4% in aqueous solution
Osmiumtetraoxide
Formule: O₄Os
MW: 254.23 g/mol
Kookpunt: 130
Smeltpunt: 40
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00011150
CAS nummer: 20816-12-0
EINECS: 244-058-7
UN: 3287
ADR: 6.1 II
Merck Index: 13,06962