(±)-2-Butaanol, anhydrous 99%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
(±)-sec-Butanol, (±)-sec-Butyl alcohol, (±)-Ethyl methyl carbinol, (±)-2-Hydroxy butane, sec-Butanol, sec-Butyl alcohol, (±)-Methyl ethyl carbinol
43315.K2
43315.AK
43315.K2EA
109
EUR
43315.K2
43315.AK
(±)-2-Butaanol, anhydrous 99%
(±)-2-Butaanol
Purity: 99%
CAS No.: 78-92-2
Molecular Formula: CH3CH2CH(OH)CH3
Formula Weight: 74.12
Flash Point: 26°(79°F)
Physical Form: Liquid
Density: 0.808
Refractive Index: 1.3970
Sensitivity: Hygroscopic
Odour: Sweet
Melting Point: -115°
Boiling Point: 98-101°
Beilstein Reference: 1718765
Merck Reference: 14,1541
UN No.: 1120
MDL No.: MFCD00004569
Formule:
H₃CCH₂CH(OH)CH₃ Kookpunt: 98…101 °C Smeltpunt: –115 °C Dichtheid: 0,808 Vlampunt: 26 °C (79 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00004569 CAS nummer: 78-92-2 EINECS: 201-158-5 UN: 1120 ADR: 3,II Merck Index: 14,1541 |