(±)-2-Pentanol for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
M-Clarity™ quality level = MQ200
8.07501.0100 8.07501.0500
8.07501.0100EA 40.7 EUR
8.07501.0100 8.07501.0500
(±)-2-Pentanol for synthesis, Sigma-Aldrich®
(±)-2-Pentanol
Formule: C₅H₁₂O
Kookpunt: 118 °C (1013 hPa)
Smeltpunt: –50 °C
Dichtheid: 0,81 g/cm³ (20 °C)
Vlampunt: 41 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004579
CAS nummer: 6032-29-7
EINECS: 227-907-6
UN: 1105
ADR: 3,III
Merck Index: 12,07258
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc37771666c375a7768be6819fbc64e80617a202&vid=cc73accd09f3b4e43eedad1fe603cf52c119ff25&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Solubility 166 g/l (20 °C)
Melting Point -50 °C
Molar Mass 88.14 g/mol
Boiling Point 119 °C (1013 hPa)
Vapor Pressure 3 hPa (20 °C)
Flash Point 41 °C
Refractive Index 1.4055 (20 °C, 589 nm)
Explosion Limit 1.2 - 10.5 %(V)
Density 0.81 g/cm3 (20 °C)
Ignition Point 330 °C
Hazard Class 3
Hazard Class Description Flammable Liquid