DL-Camphor for synthesis, Sigma-Aldrich®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: (±)-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, Camphor, 1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one, DL-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, (±)-1,7,7-Trimethyl-2-norbornanone
M-Clarity™ quality level = MQ200
8.41456.0250 8.41456.1000
8.41456.0250EA 35.5 EUR
8.41456.0250 8.41456.1000
DL-Camphor for synthesis, Sigma-Aldrich®
DL-Camphor
Formule: C₁₀H₁₆O
MW: 152.24 g/mol
Kookpunt: 204
Smeltpunt: ∼180 °C
Dichtheid: 0.992 g/cm³ (25 °C)
Vlampunt: 66
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00074738
CAS nummer: 76-22-2
EINECS: 200-945-0
UN: 2717
ADR: 4.1 III
Order Now
Specification Test Results
Solubility 1.5 g/l (20 °C)
Melting Point 180 °C
Molar Mass 152.24 g/mol
Bulk Density 800 kg/m3
Boiling Point 205 °C (1013 hPa)
Vapor Pressure 1.2 hPa (20 °C)
Flash Point 66 °C
Explosion Limit 0.6 - 4.5 %(V)
Density 0.99 g/cm3 (20 °C)
pH (H2O, 20 °C) neutral
Ignition Point 460 °C