Conductiviteit standaard (25 °C; KCl)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
42679.AP 42679.K2 42692.AP 42692.K2 42692.K7 42693.AP 42693.K2 42693.K7 42694.AP 42694.K2 42695.AP 42695.K2 42695.K7 42696.AP 42696.K2 42697.AP 42680.K2 43405.K2 42680.K7 43405.AP 42680.AP 47057.AP 47057.K2
42679.APEA 37.6 EUR
42679.AP 42679.K2 42692.AP 42692.K2 42692.K7 42693.AP 42693.K2 42693.K7 42694.AP 42694.K2 42695.AP 42695.K2 42695.K7 42696.AP 42696.K2 42697.AP 42680.K2 43405.K2 42680.K7 43405.AP 42680.AP 47057.AP 47057.K2
Conductiviteit standaard (25 °C; KCl)
Standaarden Geleidbaarheidsstandaarden
KCl conductivity standards used for calibrating conductivity cells with ranging from 718 to 100 000 µS/cm.