Kleurbakken
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
472000 472200 473100
BRND472000EA 201 EUR
BRND472000 BRND472200 BRND473100
Kleurbakken
Potjes Kleurbakjes
Soda-lime glass.
For 10 standard slides, size (76×26 mm)
Bestelinformatie: Please order staining trough, tray, and wire handle seperately.