Natriumhydroxide 1 mol/l (1 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Caustic soda
7097.1000EA 33.86 EUR
7097.1000 7097.9020 7097.9010
Natriumhydroxide 1 mol/l (1 N) in aqueous solution, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Natriumhydroxide
Formule: NaOH
MW: 40 g/mol
Kookpunt: 1390
Smeltpunt: 323
Dichtheid: 1.04 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003548
CAS nummer: 1310-73-2
EINECS: 215-185-5
UN: 3266
ADR: 8 III
Merck Index: 14,08627
Order Now
Specification Test Results
Titer (mol/l) 0.997 - 1.003
Trace Impurities (in ppm)
Chloride (Cl) max. 5
Iron (Fe) max. 0.5
Heavy Metals (as Pb) max. 1