Natriumhydroxide 2 mol/l (2 N) aqueous solution, Titripur® Volumetric, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Caustic soda
MQ100
1.09136.1000 1.09136.9025 1.09136.1003
1.09136.1000EA 23.14 EUR
1.09136.1000 1.09136.9025 1.09136.1003
Natriumhydroxide 2 mol/l (2 N) aqueous solution, Titripur® Volumetric, Supelco®
Natriumhydroxide
Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.
Formule: NaOH
MW: 40 g/mol
Kookpunt: 1390
Smeltpunt: 323
Dichtheid: ≤1.11 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003548
CAS nummer: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 II
Merck Index: 14,08627
Order Now
Specification Test Results
Form liquid
Amount-of-substance concentration 1.990 - 2.010
Measurement uncertainty +/- 0.006
Traceability NIST SRM
The concentration is determined by volumetric titration and refers to 20°C.