Natriumthiosufaat standardised solution 0.1 N in aqueous solution
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate
35645.K2 35645.M4
35645.K2EA 35.7 EUR
35645.K2 35645.M4
Natriumthiosufaat standardised solution 0.1 N in aqueous solution
Natriumthiosufaat
CAS No.: 7772-98-7 Molecular Formula: Na2O3S2 Formula Weight: 158.10 Physical Form: Liquid Odour: Odourless MDL No.: MFCD00003499
Formule: Na₂S₂O₃
MW: 158.11 g/mol
Kookpunt: 100
Smeltpunt: 48
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003499
CAS nummer: 7772-98-7
EINECS: 231-867-5
Merck Index: 12,08844