Zwavelzuur standardised solution 1.0 N in aqueous solution
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
35655.K2 35655.M4
35655.K2EA 31.1 EUR
35655.K2 35655.M4
Zwavelzuur standardised solution 1.0 N in aqueous solution
Zwavelzuur
Molecular Formula: H2SO4 Formula Weight: 98.08 Physical Form: Liquid Appearance: Colourless to pale yellow UN No.: 3264 MDL No.: MFCD00064589
Formule: H₂SO₄
Dichtheid: 1,01 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064589
CAS nummer: 7664-93-9
EINECS: 231-639-5
UN: 3264
ADR: 8,III
Order Now
Specificaties
Environmentally Preferable