Zilvernitraat 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM USP, Reag. Ph. Eur. volumetric solution
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Lunar caustic, Silver mononitrate, Nitric acid silver salt
30472.297EA 111 EUR
30472.297 30472.322 30472.402 VWRS30472.323 30472.290
Zilvernitraat 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM USP, Reag. Ph. Eur. volumetric solution
Zilvernitraat
Formule: AgNO₃
MW: 169.87 g/mol
Kookpunt: 444
Smeltpunt: 210
Dichtheid: 1,01 g/cm3
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003414
CAS nummer: 7761-88-8
EINECS: 231-853-9
UN: 3264
ADR: 8 III
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Titer (20°C; real value 0.2 % accuracy) 0.0998 - 0.1002 mol/l
Conforms to Reag. Ph.Eur. Passes test
Conforms to USP Passes test
NIST traceable Confirmed