Citric acid monohydrate 99,5
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid
22869.22 22869.36 22869.A1
22869.22EA 26.7 EUR
22869.22 22869.36 22869.A1
Citric acid monohydrate 99,5
Citric acid monohydrate
Purity: 99.5+% CAS No.: 5949-29-1 Molecular Formula: C6H8O7.H2O Formula Weight: 210.14 (192.13anhy) Physical Form: Granular Density: 1.542 Appearance: White Odour: Odourless Merck Reference: 14,2326 MDL No.: MFCD00149972
Formule: C₆H₈O₇.H₂O
MW: 210.14 g/mol
Smeltpunt: 135…152 °C
Dichtheid: 1.542
Vlampunt: 173.9 °C (gesloten beker)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149972
CAS nummer: 5949-29-1
EINECS: 201-069-1
Merck Index: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3160681f96abd248f56672a96b72cc5f6e9d89a2&vid=c4db52c32c630d2c9876e60fc3e9e70a117ea0d8&prid=web&clid=SAPDAM