Citric acid monohydrate 99,0-102,0 ACS
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid
36665.22 36665.36 36665.A3
36665.22EA 33.6 EUR
36665.22 36665.36 36665.A3
Citric acid monohydrate 99,0-102,0 ACS
Citric acid monohydrate
Purity: 99.0-102.0% CAS No.: 5949-29-1 Molecular Formula: C6H8O7.H2O Formula Weight: 210.14 (192.13anhy) Physical Form: Solid Density: 1.542 Appearance: White Odour: Odourless Merck Reference: 14,2326 MDL No.: MFCD00149972
Formule: HOC(COOH)(CH₂COOH)₂·H₂O
MW: 210.14 g/mol
Smeltpunt: 135…152 °C
Dichtheid: 1.542
Vlampunt: 173.9 °C (gesloten beker)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149972
CAS nummer: 5949-29-1
EINECS: 201-069-1
Merck Index: 14,2326
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3160681f96abd248f56672a96b72cc5f6e9d89a2&vid=c4db52c32c630d2c9876e60fc3e9e70a117ea0d8&prid=web&clid=SAPDAM