Lithium hydroxide monohydrate ≥98.5%, Purum, Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
13020-1KG 13020-6X1KG
13020-1KGEA 234 EUR
13020-1KG 13020-6X1KG
Lithium hydroxide monohydrate ≥98.5%, Purum, Fluka™
Lithium hydroxide monohydrate
  • ≥95% for general laboratory use.
Formule: LiOH·1H₂O
Kookpunt: 920 °C (1013 hPa)
Smeltpunt: 423 °C
Dichtheid: 1,830 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149772
CAS nummer: 1310-66-3
EINECS: 215-183-4
UN: 2680
ADR: 8,II
Merck Index: 13,05556
Order Now
Specification Test Results
Assay Min. 98.5 %
Assay of Li₂CO₃ Max. 1 %
Calcium (Ca) Max. 0.05 %
Iron (Fe) Max. 0.001 %
Potassium (K) Max. 0.02 %
Sodium (Na) Max. 0.02 %
Chloride (Cl) Max. 0.05 %
Sulfate (SO₄) Max. 0.05 %