Tetrasodium diphosphate decahydrate
Leverancier: Roth Carl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Sodium pyrophosphate decahydrate, tetra-Sodium pyrophosphate decahydrate, Sodium pyrophosphate tetrabasic decahydrate, TSPP
0269.1 0269.2 0269.3
ROTH0269.1EA 0 EUR
ROTH0269.1 ROTH0269.2 ROTH0269.3
Tetrasodium diphosphate decahydrate
Tetrasodium diphosphate decahydrate
Formule: Na₄P₂O₇·10H₂O
MW: 446.06 g/mol
Smeltpunt: 990
Dichtheid: 1.82 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149200
CAS nummer: 13472-36-1
EINECS: 231-767-1