Natriumdiwaterstoffosfaat dihydraat, EMPROVE® ESSENTIAL BP, E339, Ph. Eur., USP, JPE suitable for use as excipient, SAFC®
Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Phosphoric acid (sodium hydrate), Sodium phosphate monobasic dihydrate, Sodium dihydrogen orthophosphate dihydrate
1.06345.1000 1.06345.5000 1.06345.9026 1.06345.9011
1.06345.1000EA 141 EUR
1.06345.1000 1.06345.5000 1.06345.9026 1.06345.9011
Natriumdiwaterstoffosfaat dihydraat, EMPROVE® ESSENTIAL BP, E339, Ph. Eur., USP, JPE suitable for use as excipient, SAFC®
Natriumdiwaterstoffosfaat dihydraat
Formule: NaH₂PO₄·2H₂O
MW: 156.01 g/mol
Kookpunt: ∼650 °C (1013 hPa)
Smeltpunt: 60
Dichtheid: 1.91 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149209
CAS nummer: 13472-35-0
EINECS: 231-449-2
Bereik: 01-2119489796-13
Merck Index: 12,08806