Calciumnitraat tetrahydraat 99.0-103.0% ACS
Leverancier: Thermo Fisher Scientific
Ratings:
Avg. Ratings: 1.0 (1 Review)
|
Gevaar
|
12364.36
12364.A3
12364.36EA
46.6
EUR
12364.36
12364.A3
Calciumnitraat tetrahydraat 99.0-103.0% ACS
Calciumnitraat tetrahydraat
Purity: 99.0-103.0%
CAS No.: 13477-34-4
Molecular Formula: Ca(NO3)2.4H2O
Formula Weight: 236.15 (164.09anhy)
Physical Form: Crystalline
Density: 1.82
Melting Point: 42°
Boiling Point: 132° dec.
Solubility: Soluble in water, alcohol, and acetone
Merck Reference: 14,1682
UN No.: 1454
MDL No.: MFCD00149604
Manufacturing of other nitrates, fertilizers, matches
Formule:
Ca(NO₃)₂·4H₂O Kookpunt: 132 °C (ontbinding) Smeltpunt: 42 °C Dichtheid: 1,82 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00149604 CAS nummer: 13477-34-4 EINECS: 233-332-1 UN: 1454 ADR: 5.1,III Merck Index: 14,1682 |