Calciumnitraat tetrahydraat 97+% (by complexometric analysis)
Leverancier: Thermo Fisher Scientific
Ratings:
Avg. Ratings: 1.0 (1 Review)
Total Ratings: 1
Avg. Ratings: 1.0 out of 5
Was this review helpful? Yes No
Yes: 0 No: 1 
pharmaceutical system / laboratory technician / grenoble, france
No, I didn't recommend
for R&D purpose only ! not mentionned in the description ! / pour utilisation de R&D seulement pas précisé au moment de l'achat !
Gevaar
A16645.0E A16645.22 A16645.36
A16645.0EEA 132 EUR
A16645.0E A16645.22 A16645.36
Calciumnitraat tetrahydraat 97+% (by complexometric analysis)
Calciumnitraat tetrahydraat
Purity: 99% CAS No.: 13477-34-4 Molecular Formula: Ca(NO3)2.4H2O Formula Weight: 236.15 (164.09anhy) Odour: Odourless Melting Point: 43-45° Boiling Point: 132° dec. Merck Reference: 14,1682 UN No.: 1454 MDL No.: MFCD00149604
Formule: Ca(NO₃)₂·4H₂O
MW: 236.15 g/mol
Kookpunt: 132 °C (ontbinding)
Smeltpunt: 43…45 °C
Dichtheid: 1.820
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149604
CAS nummer: 13477-34-4
EINECS: 233-332-1
UN: 1454
ADR: 5.1 III
Merck Index: 13,01687