Magnesium nitrate hexahydrate 98
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
A10329.0B A10329.0I A10329.30
A10329.0BEA 56.6 EUR
A10329.0B A10329.0I A10329.30
Magnesium nitrate hexahydrate 98
Magnesium nitrate hexahydrate
Purity: 98% CAS No.: 13446-18-9 Molecular Formula: Mg(NO3)2.6H2O Formula Weight: 256.41 (148.32anhy) Melting Point: 89° Boiling Point: 330° dec. Merck Reference: 14,5674 UN No.: 1474 MDL No.: MFCD00149779
Formule: Mg(NO₃)₂·6H₂O
MW: 256.41 g/mol
Kookpunt: 330 °C (ontbinding)
Smeltpunt: 89
Dichtheid: 1.46
Vlampunt: 93,3
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149779
CAS nummer: 13446-18-9
EINECS: 233-826-7
UN: 1474
ADR: 5.1 III
Merck Index: 13,05697