Aluminium chloride hexahydrate ≥99.9995% (metals basis), Puratronic®
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Aluminum chloride hexahydrate
10622.06
10622.18
10622.30
10622.06EA
25.2
EUR
10622.06
10622.18
10622.30
Aluminium chloride hexahydrate ≥99.9995% (metals basis), Puratronic®
Aluminium chloride hexahydrate
Purity: 99.9995%
CAS No.: 7784-13-6
Molecular Formula: AlCl3.6H2O
Formula Weight: 241.43 (133.34anhy)
Physical Form: Crystalline Aggregates
Density: 2.398
Appearance: White
Odour: Odourless
pH: 2.5
Melting Point: 100° dec.
Merck Reference: 14,337
UN No.: 3260
MDL No.: MFCD00149134
Formule:
AlCl₃·6H₂O MW: 241.43 g/mol Smeltpunt: 100 °C (ontbinding) Dichtheid: 2.398 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00149134 CAS nummer: 7784-13-6 EINECS: 231-208-1 UN: 3260 ADR: 8 III Merck Index: 14,337 |