Tetrahydrofuraan, ultra-low water content ≥99.8% (by GC, corrected for water content), BakerDRY™ for organic synthesis, J.T.Baker®
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Butylene oxide, THF, Diethylene oxide
Packaged under argon.
9446-12EA 562.74 EUR
9446-12 9446-10
Tetrahydrofuraan, ultra-low water content ≥99.8% (by GC, corrected for water content), BakerDRY™ for organic synthesis, J.T.Baker®
Tetrahydrofuraan
Specifically manufactured for use in organic, organometallic, and oligonucleotide synthesis requiring low water solvents.
Formule: C₄H₈O
MW: 72.11 g/mol
Kookpunt: 66
Smeltpunt: –108.5 °C
Dichtheid: 0.888 g/cm³ (20 °C)
Vlampunt: –21.5 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005356
CAS nummer: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3 II
Bereik: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use = 99.8 %
ACS - Assay (by GC, corrected for water) ≥99.8 %
ACS - Color (APHA) ≤10
ACS - Peroxide (as H₂O₂) (at time of manufacture) ≤0.015 %
ACS - Residue after Evaporation ≤1 ppm
ACS - Water (by KF, coulometric) ≤10.0 ppm
Packaged under Argon