1-Bromopentadecane ≥97%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
L07618.09 L07618.18
L07618.09EA 49.3 EUR
L07618.09 L07618.18
1-Bromopentadecane ≥97%
1-Bromopentadecane
Purity: 97% CAS No.: 629-72-1 Molecular Formula: CH3(CH2)14Br Formula Weight: 291.32 Flash Point: 110°(230°F) Refractive Index: 1.4608 Melting Point: 18-19° Boiling Point: 159-160°/5mm Beilstein Reference: 1744553 MDL No.: MFCD00000229
Formule: BrH₂C(CH₂)₁₃CH₃
Kookpunt: 159…160 °C (5 mmHg)
Smeltpunt: 18…19 °C
Dichtheid: 1,005
Vlampunt: >110 °C (230 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000229
CAS nummer: 629-72-1
EINECS: 211-104-2