Afdichtfilm en -folie voor PP microplaten
Leverancier: EPPENDORF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0030127.889 0030127.870
713-0007EA 142 EUR
713-0007 EPPE0030127.870
Afdichtfilm en -folie voor PP microplaten
Afdichtingen Afdichtingen van microtiterplaten
Secure your samples with Eppendorf® Microplate self-adhesive film and foil, perfect for incubation and short-term storage. Films protect from punctures while aluminium foils shield light-sensitive samples with easy sealing.
  • Storage films are transparent and provide protection against unwanted punctures
  • Storage foils are made of aluminium, can be pierced, and protect light-sensitive samples
  • Storage film and foil glued seals for simple and dependable sealing during sample storage
For long-term storage, we recommend the use of an Eppendorf heat sealing film.