Glasvezelvoorfilters, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
DS0281-5000 DS0281-7500 DS0281-9000
513-2811EA 125 EUR
513-2811 NALGDS0281-7500 513-2812
Glasvezelvoorfilters, Nalgene®
Filters Glasvezelfilters
Increase filtration speed and throughput of highly viscous or heavily particulated solutions by using these glass fiber prefilter discs in combination with any Nalgene™ MF75 filter units or bottle top filters.