Exsiccatorplaten
Leverancier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
66015 66025 66030 65965 65975 65980
BRND66015EA 14.3 EUR
BRND66015 BRND66025 BRND66030 BRND65965 BRND65975 BRND65980
Exsiccatorplaten
Exsiccatoren Exsiccatorplaten
Desiccator plates for glass and plastic desiccators.
  • Central hole measures approximately 20 mm diameter
  • Small perforations measure approximately 5 mm diameter
Order Now
Specificaties
Environmentally Preferable