Acetonitril, extra dry 99.9+%, AcroSeal®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
326811000 326810010 326810025
326811000.EA 78.2 EUR
326811000. 326810010. 326810025.
Acetonitril, extra dry 99.9+%, AcroSeal®
Acetonitril
Formule: H₃CCN
Kookpunt: 81…82 °C (760 mmHg)
Smeltpunt: –46 °C
Dichtheid: 0,781 g/cm³
Vlampunt: 2 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001878
CAS nummer: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
Bereik: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.001 % (Coulometric)
GC ≥99.90 %
Residue after evaporation ≤0.0005 %
Acidity (CH3COOH) ≤0.002 %
Acetamide ≤10 ppm