Acetonitril ≥99.7%, Environmental Grade
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Purity: 99.7+%
CAS No.: 75-05-8
Molecular Formula: CH3CN
Formula Weight: 41.05
Flash Point: 5°(41°F)
Physical Form: Liquid
Density: 0.786
Refractive Index: 1.3440
Odour: Pungent
Melting Point: -48° to -45°
Boiling Point: 80-82°
Beilstein Reference: 741857
Merck Reference: 14,70
UN No.: 1648
MDL No.: MFCD00001878
Suitable for environmental, pesticide residue, and GC analysis
Formule:
H₃CCN Kookpunt: 80…82 °C Smeltpunt: –48 °C…–45 °C Dichtheid: 0,786 Vlampunt: 5 °C (41 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00001878 CAS nummer: 75-05-8 EINECS: 200-835-2 UN: 1648 ADR: 3,II Bereik: 01-2119471307-38 Merck Index: 14,70 |