Slangaansluitingen, recht, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6150-0010 6150-0020
NALG6150-0010EA 42.2 EUR
NALG6150-0010 NALG6150-0020
Slangaansluitingen, recht, Nalgene®
Fittingen en connectoren
HDPE, translucent.
  • Good chemical resistance
  • Good temperature resistance
Two tapered tubulations fit tightly together with male/female connection, allowing for rapid joining or separation of apparatus lines.