α-Chloro-4-(trifluoromethoxy)toluene

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 1-(Chloromethyl)-4-(trifluoromethoxy)benzene, 4-(Trifluoromethoxy)benzyl chloride, p-(Trifluoromethoxy)benzyl chloride, 1-(Chlormethyl)-4-(trifluormethoxy)benzene

TL00386DA TL00386EA
MAYBTL00386DAEA 0 EUR
MAYBTL00386DA MAYBTL00386EA
α-Chloro-4-(trifluoromethoxy)toluene
α-Chloro-4-(trifluoromethoxy)toluene
Formule: C₈H₆ClF₃O
MW: 210.58
MDL Nummer: MFCD00052326
CAS nummer: 65796-00-1
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR