3',4'-Difluoroacetophenone

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 1-(3,4-Difluorophenyl)ethan-1-one, 1-(3,4-Difluorophenyl)ethanone

BTB06035.10 BTB06035.50 BTB06035.1
MAYBBTB06035.10EA 0 EUR
MAYBBTB06035.10 MAYBBTB06035.50 MAYBBTB06035.1
3',4'-Difluoroacetophenone
3',4'-Difluoroacetophenone
Formule: C₈H₆F₂O
MW: 156.13 g/mol
Kookpunt: 207
Smeltpunt: 20
Dichtheid: 1.24 g/cm³ (20 °C)
MDL Nummer: MFCD00009891
CAS nummer: 369-33-5
EINECS: 206-717-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR