5-Chloro-o-anisidine hydrochloride
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SB01226.1 SB01226.10 SB01226.50
MAYBSB01226.1EA 0 EUR
MAYBSB01226.1 MAYBSB01226.10 MAYBSB01226.50
5-Chloro-o-anisidine hydrochloride
5-Chloro-o-anisidine hydrochloride
Formule: C₇H₉Cl₂NO
MDL Nummer: MFCD00035488
CAS nummer: 4274-03-7
EINECS: 224-272-7
UN: 2811
ADR: 6.1,III