4'-Butylacetophenone

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BTB09784.10 BTB09784.50 BTB09784.1
MAYBBTB09784.10EA 0 EUR
MAYBBTB09784.10 MAYBBTB09784.50 MAYBBTB09784.1
4'-Butylacetophenone
4'-Butylacetophenone
Formule: C₁₂H₁₆O
MDL Nummer: MFCD00017500
CAS nummer: 37920-25-5
EINECS: 253-715-7

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR