1-Chloor-2-propanol ≥75% (remainder mainly 2-chloro-1-propanol), Technical Grade
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: 1-Chloropropan-2-ol, Propylene chlorohydrin, Chloropropanol
B22686.14 B22686.22
B22686.14EA 45.3 EUR
B22686.14 B22686.22
1-Chloor-2-propanol ≥75% (remainder mainly 2-chloro-1-propanol), Technical Grade
1-Chloor-2-propanol
Purity: 75% CAS No.: 127-00-4 Molecular Formula: CH3CH(OH)CH2Cl Formula Weight: 94.54 Flash Point: 51°(123°F) Refractive Index: 1.4370 Boiling Point: 126-127° Beilstein Reference: 773653 Merck Reference: 14,7850 UN No.: 2929 MDL No.: MFCD00004530
Formule: C₃H₇ClO
MW: 94.54 g/mol
Kookpunt: 126…127 °C
Dichtheid: 1.111
Vlampunt: 51 °C (123 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004530
CAS nummer: 127-00-4
EINECS: 204-819-6
UN: 2611
ADR: 6.1 II
Merck Index: 14,7850