5'-Chloro-2'-hydroxyacetophenone

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 1-(5-Chloro-2-hydroxyphenyl)ethan-1-one, 1-(5-Chloro-2-hydroxyphenyl)ethanone

BTB07442.1 BTB07442.10
MAYBBTB07442.1EA 0 EUR
MAYBBTB07442.1 MAYBBTB07442.10
5'-Chloro-2'-hydroxyacetophenone
5'-Chloro-2'-hydroxyacetophenone
Formule: C₈H₇ClO₂
MDL Nummer: MFCD00067788
CAS nummer: 1450-74-4
EINECS: 215-916-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR