4'-Decylacetophenone

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 1-(4-Decylphenyl)ethanone, p-Decylacetophenone

BTB09790.1 BTB09790.10
MAYBBTB09790.1EA 0 EUR
MAYBBTB09790.1 MAYBBTB09790.10
4'-Decylacetophenone
4'-Decylacetophenone
Formule: C₁₈H₂₈O
MDL Nummer: MFCD00173754
CAS nummer: 37593-06-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR