2-Chlorobenzimidazole 97%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
2-Chloro-1H-benzo[d]imidazole, 2-Chloro-1H-benzimidazole
291960250
AC29196.1
AC29196.10
AC29196.50
ACRO291960250EA
78.4
EUR
ACRO291960250
MAYBAC29196.1
MAYBAC29196.10
MAYBAC29196.50
2-Chlorobenzimidazole 97%
2-Chlorobenzimidazole
Formule:
C₇H₅ClN₂ Smeltpunt: 200 °C |
MDL Nummer:
MFCD00051944 CAS nummer: 4857-06-1 EINECS: 225-453-3 |
Specification Test Results
Appearance | Beige to brown Crystalline powder |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...