Trisodium citrate, anhydrous 99% (by titrimetric analysis)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Sodium Citrate, Sodium citrate tribasic, Citric acid trisodium salt, Sodium 2-hydroxypropane-1,2,3-tricarboxylate
45556.A1 45556.30 45556.A7
45556.A1EA 114 EUR
45556.A1 45556.30 45556.A7
Trisodium citrate, anhydrous 99% (by titrimetric analysis)
Trisodium citrate
Trisodium citrate is used in food additives, beverages, instant drinks, pharmaceuticals and detergents. It is also used as buffering agent, acidity regulator, sequestering or emulsifying agent.
It is an effective agent for removal of carbonate scale from boilers and for cleaning automobile radiators. In the medical field, it is used as an osmotic laxative and an active component of the WHO oral rehydration solution. It is also employed to control acidity in some substances such as gelatin desserts. Further, it is used as a flavour or as a preservative. In addition to this, it is used for the preservation of blood in blood banks.
Formule: C₆H₅Na₃O₇
MW: 258.07 g/mol
Smeltpunt: 300 °C
Dichtheid: 1.815 g/cm³ (20 °C)
Vlampunt: 290
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00012462
CAS nummer: 68-04-2
EINECS: 200-675-3
Merck Index: 14,8602
Order Now
Specification Test Results
Assay (Titration) >98.5%