4-(Bromomethyl)benzenesulphonyl chloride 95%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
4-(Bromomethyl)benzene-1-sulphonyl chloride
A10120.06
A10120.14
A10120.06EA
86.8
EUR
A10120.06
A10120.14
4-(Bromomethyl)benzenesulphonyl chloride 95%
4-(Bromomethyl)benzenesulphonyl chloride
Purity: 95%
CAS No.: 66176-39-4
Molecular Formula: C7H6BrClO2S
Formula Weight: 269.55
Melting Point: 72-75°
Beilstein Reference: 2364328
UN No.: 3261
MDL No.: MFCD00156129
Formule:
C₇H₆BrClO₂S MW: 269.55 Smeltpunt: 72…75 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00156129 CAS nummer: 66176-39-4 UN: 3261 ADR: 8 II |